Learn More
Eppendorf™ 15mL Steel Sleeve
Brand:  Eppendorf™ 5702707007
178.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11532235
  • 178.71 EUR / Pack of 10
Specifications
Sleeve
Rotor F–35-30-17
Product Title
Eppendorf™ 15mL Steel Sleeve > For Rotor F-35-30-17