Learn More
Bochem™ 18/10 Stainless Mini Funnels
Brand:  Bochem™ 8857
Product Code. 12671895
  • 4.45 EUR / Each
Estimated Shipment: 09-05-2025
Specifications
8/29mm
Stainless Steel (18/10)
50 mm
Mini Funnel
Product Title
Bochem™ 18/10 Stainless Mini Funnels > Diameter: 0.275mm