Learn More
Fisher Chemical 18LT METHYLATED SPIRIT (IDA 99)99%V/V ANHYDROUS
Brand:  Fisher Chemical M/4450/30
18LT METHYLATED SPIRIT (IDA 99)99%V/V ANHYDROUS
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Product Code. 11402884
  • 220.00 EUR / 18 L
Videos
Safety and Handling
Show More
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2533170 Certificate TypeCertificate of Analysis Date03/04/2025 Catalog NumberM/4450/30,M/4450/27,M/4450/21
Lot Number2532072 Certificate TypeCertificate of Analysis Date26/03/2025 Catalog NumberM/4450/30,M/4450/27,M/4450/21
Lot Number2530833 Certificate TypeCertificate of Analysis Date18/03/2025 Catalog NumberM/4450/30,M/4450/27,M/4450/21
Lot Number2530830 Certificate TypeCertificate of Analysis Date18/03/2025 Catalog NumberM/4450/30,M/4450/27,M/4450/21
Lot Number2530836 Certificate TypeCertificate of Analysis Date16/03/2025 Catalog Number
Product Title
Fisher Chemical 18LT METHYLATED SPIRIT (IDA 99)99%V/V ANHYDROUS >