missing translation for 'onlineSavingsMsg'
Learn More
2-(1-Piperidinyl)aniline, 98%, Thermo Scientific Chemicals Product Code.: 11404394

2-(1-Piperidinyl)aniline, 98%, Thermo Scientific Chemicals

Product Code. 11404394
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11404394

Brand: Thermo Scientific Alfa Aesar A13073.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39643-31-7
Molecular Formula C11H16N2
Molecular Weight (g/mol) 176.26
MDL Number MFCD00047467
InChI Key OYECAJPUPWFCSL-UHFFFAOYSA-N
Synonym 2-piperidinoaniline, 2-piperidin-1-yl aniline, 2-piperidin-1-yl-phenylamine, benzenamine, 2-1-piperidinyl, n-2-aminophenyl piperidine, 2-1-piperidinyl aniline, 2-1-piperidino aniline, 1-2-aminophenyl piperidine, 2-piperidylphenylamine, 2-piperdinoaniline
PubChem CID 458795
IUPAC Name 2-piperidin-1-ylaniline
SMILES NC1=CC=CC=C1N1CCCCC1

Specifications

Melting Point 44°C to 47°C
Boiling Point 96°C to 98°C (1 mmHg)
Quantity 100 g
UN Number UN2811
Beilstein 151856
Formula Weight 176.26
Percent Purity 98%
Chemical Name or Material 2-(1-Piperidinyl)aniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-(1-Piperidinyl)aniline, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.