missing translation for 'onlineSavingsMsg'
Learn More
2,2'-Diaminodiphenyl disulfide, 97%, Thermo Scientific Chemicals Product Code.: 15402168

2,2'-Diaminodiphenyl disulfide, 97%, Thermo Scientific Chemicals

Product Code. 15402168
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 15402168

Brand: Thermo Scientific Alfa Aesar H55807.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2,2'-Diaminodiphenyl disulfide, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′-Diaminodiphenyl disulfide, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a primary and secondary intermediate.

Solubility
Soluble in hot methanol- almost transparent.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1141-88-4
Molecular Formula C12H12N2S2
Molecular Weight (g/mol) 248.36
MDL Number MFCD00007703
InChI Key YYYOQURZQWIILK-UHFFFAOYSA-N
Synonym 2,2'-dithiodianiline, benzenamine, 2,2'-dithiobis, intramine, 2,2'-disulfanediyldianiline, 2-aminophenyl disulfide, 2,2'-diaminodiphenyldisulfide, 2,2'-diaminodiphenyl disulfide, 2-2-aminophenyl disulfanyl aniline, aniline, 2,2'-dithiodi, bis o-aminophenyl disulfideShow MoreShow Less
PubChem CID 14358
IUPAC Name 2-[(2-aminophenyl)disulfanyl]aniline
SMILES NC1=CC=CC=C1SSC1=CC=CC=C1N

Specifications

Melting Point 91°C to 92°C
Quantity 25 g
Beilstein 914032
Solubility Information Soluble in hot methanol- almost transparent.
Formula Weight 248.36
Percent Purity 97%
Chemical Name or Material 2,2'-Diaminodiphenyl disulfide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT13I016 Certificate TypeCertificate of Analysis Date19/07/2022 Catalog NumberH55807.06,H55807.14
Lot NumberY03H017 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberH55807.06,H55807.14
Lot Number81202155 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH55807.06,H55807.14
Lot NumberP10G024 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH55807.06,H55807.14
Lot Number5003R22U Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH55807.06,H55807.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,2'-Diaminodiphenyl disulfide, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.