Learn More
2-(2-Naphthyl)-4-(5,6,7,8-tetrahydro-2-naphthyl)thiazole, 97%, Thermo Scientific™ Product Code.: 15434216
2-(2-Naphthyl)-4-(5,6,7,8-tetrahydro-2-naphthyl)thiazole, 97%, Thermo Scientific™
Product Code. 15434216
10 g
Click to view available options
Quantity:
2 g
10 g
Product Code. 15434216
Brand: Thermo Scientific Alfa Aesar H58405.09
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1421261-80-4
Molecular Formula C23H19NS
Molecular Weight (g/mol) 341.472
MDL Number MFCD22682973
InChI Key ISODJAPESPIWDI-UHFFFAOYSA-N
Synonym 2-naphthalen-2-yl-4-5,6,7,8-tetrahydronaphthalen-2-yl-1,3-thiazole, 2-2-naphthyl-4-5,6,7,8-tetrahydro-2-naphthyl thiazole
PubChem CID 97030349
IUPAC Name 2-naphthalen-2-yl-4-(5,6,7,8-tetrahydronaphthalen-2-yl)-1,3-thiazole
SMILES C1CCC2=C(C1)C=CC(=C2)C3=CSC(=N3)C4=CC5=CC=CC=C5C=C4
Specifications
Quantity 10 g
Formula Weight 341.47
Percent Purity 97%
Chemical Name or Material 2-(2-Naphthyl)-4-(5,6,7,8-tetrahydro-2-naphthyl)thiazole
Product Content Correction
Product Title
2-(2-Naphthyl)-4-(5,6,7,8-tetrahydro-2-naphthyl)thiazole, 97%, Thermo Scientific™ >