missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Thienyl)ethyl isocyanate, 95+%, Thermo Scientific™

33.80 EUR - 386.00 EUR

Chemical Identifiers

CAS 58749-51-2
Molecular Formula C7H7NOS
Molecular Weight (g/mol) 153.199
InChI Key KAJZMNLREUEZSH-UHFFFAOYSA-N
PubChem CID 2782179
IUPAC Name 2-(2-isocyanatoethyl)thiophene
SMILES C1=CSC(=C1)CCN=C=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10713263
SDS View Documents
Thermo Scientific Maybridge
RF05240CB
250 mg
33.80 EUR
250mg
Please to purchase this item. Need a web account? Register with us today!
10580353
SDS View Documents
Thermo Scientific Maybridge
RF05240DA
1 g
90.20 EUR
1g
Please to purchase this item. Need a web account? Register with us today!
10737305
SDS View Documents
Thermo Scientific Maybridge
RF05240EA
10 g
386.00 EUR
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications
Show More
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-(2-Thienyl)ethyl isocyanate, 95+%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.