missing translation for 'onlineSavingsMsg'
Learn More
2,3,5-Tri-O-benzyl-D-ribofuranose, 98%, Thermo Scientific Chemicals Product Code.: 15406457

2,3,5-Tri-O-benzyl-D-ribofuranose, 98%, Thermo Scientific Chemicals

Product Code. 15406457
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15406457

Brand: Thermo Scientific Alfa Aesar H33605.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54623-25-5
Molecular Formula C26H28O5
MDL Number MFCD03425640

Specifications

Quantity 1 g
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 420.5
Percent Purity 98%
Chemical Name or Material 2,3,5-Tri-O-benzyl-D-ribofuranose

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10199740 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH33605.03,H33605.06
Lot Number10166985 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH33605.03,H33605.06
Lot Number10192195 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH33605.03,H33605.06
Lot Number10204423 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH33605.03,H33605.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,3,5-Tri-O-benzyl-D-ribofuranose, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.