missing translation for 'onlineSavingsMsg'
Learn More

2,3,6,2',3',4',6'-Hepta-O-acetyl-alpha-D-lactosyl bromide, Thermo Scientific Chemicals

39.80 EUR - 304.52 EUR

Chemical Identifiers

CAS 4753-07-5
Molecular Formula C26H35BrO17
Molecular Weight (g/mol) 699.45
MDL Number MFCD00153593
InChI Key NLFHLQWXGDPOME-UHFFFAOYNA-N
Synonym bromo heptaacetyl-d-lactoside, stabilized with calcium carbonate, bromo heptaacetyl-d-lactoside, 2,3,6,2 ,3 ,4 ,6-hepta-o-acetyl-a-d-cellobiosyl bromide, 2,2',3,3',4',6,6'-hepta-o-acetyl-alpha-d-lactosyl bromide, 2,3,6,2',3',4',6'-hepta-o-acetyl-1-bromo-1-deoxy-alpha-d-lactopyranose min, 2r,3s,4s,5r,6s-3,4,5-tris acetyloxy-6-2r,3r,4s,5r,6r-4,5-bis acetyloxy-2-acetyloxy methyl-6-bromooxan-3-yl oxy oxan-2-yl methyl acetate, 2-o,3-o,6-o-triacetyl-4-o-2-o,3-o,4-o,6-o-tetraacetyl-beta-d-galactopyranosyl-alpha-d-glucopyranosyl bromide, acetobromo-, a-d-lactoside, bromo 4-o-2 ,3,4,6-tetra-o-acetyl-, a-d-galactopyranosyl-2,3,6-tri-o-acetyl-, a-d-glucopyranoside,
PubChem CID 11136238
IUPAC Name [(2R,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-[(2R,3R,4S,5R,6R)-4,5-diacetyloxy-2-(acetyloxymethyl)-6-bromooxan-3-yl]oxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC2C(OC(C(C2OC(=O)C)OC(=O)C)Br)COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15460567
SDS View Documents
Thermo Scientific Alfa Aesar
J66385.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15470567
SDS View Documents
Thermo Scientific Alfa Aesar
J66385.06
5 g
171.86 EUR
5g
Please to purchase this item. Need a web account? Register with us today!
15480567
SDS View Documents
Thermo Scientific Alfa Aesar
J66385.09
10 g
304.52 EUR
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in chloroform

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Store at -20°C; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,3,6,2',3',4',6'-Hepta-O-acetyl-alpha-D-lactosyl bromide, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.