missing translation for 'onlineSavingsMsg'
Learn More
2,3-O-Isopropylidene-alpha,beta-D-ribofuranose, Thermo Scientific Chemicals Product Code.: 11362827

2,3-O-Isopropylidene-alpha,beta-D-ribofuranose, Thermo Scientific Chemicals

Product Code. 11362827
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11362827

Brand: Thermo Scientific Alfa Aesar L15988.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

2,3-O-Isopropylidene-alpha,beta-D-ribofuranose is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-O-Isopropylidene-alpha,beta-D-ribofuranose is used as a pharmaceutical intermediate.

Solubility
Soluble in acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13199-25-2
Molecular Formula C8H14O5
MDL Number MFCD01075718

Specifications

Quantity 5 g
Solubility Information Soluble in acetone.
Optical Rotation −27° (c=1 in Chloroform)
Formula Weight 190.2
Chemical Name or Material 2,3-O-Isopropylidene-alpha,beta-D-ribofuranose

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10208169 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL15988.03,L15988.06
Lot NumberL03Y006 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL15988.03,L15988.06
Lot NumberH10X009 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL15988.03,L15988.06
Lot Number10147857 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL15988.03,L15988.06
Lot NumberJ14U011 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL15988.03,L15988.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,3-O-Isopropylidene-alpha,beta-D-ribofuranose, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.