missing translation for 'onlineSavingsMsg'
Learn More
2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol, 96%, Thermo Scientific Chemicals Product Code.: 11426966

2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol, 96%, Thermo Scientific Chemicals

Product Code. 11426966
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11426966

Brand: Thermo Scientific Alfa Aesar B24048.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 722-92-9
Molecular Formula C9H7F6NO
Molecular Weight (g/mol) 259.151
MDL Number MFCD00039258
InChI Key TZEJXCIGVMTMDY-UHFFFAOYSA-N
Synonym 2-4-aminophenyl-1,1,1,3,3,3-hexafluoropropan-2-ol, 4-hexafluoro-2-hydroxyisopropyl aniline, 2-4-aminophenyl-1,1,1,3,3,3-hexafluoro-2-propanol, 2-4-aminophenyl hexafluoro-2-propanol, 2-4-amino-phenyl-1,1,1,3,3,3-hexafluoro-propan-2-ol, benzenemethanol,4-amino-a,a-bis trifluoromethyl, enamine_005408, 4-hexafluoro-2-hydroxy-2-propyl aniline, 4-hexafluoro-2-hydroxy-2-propyl-aniline, 2-4-aminophenyl hexafluoropropan-2-ol
PubChem CID 577801
IUPAC Name 2-(4-aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol
SMILES C1=CC(=CC=C1C(C(F)(F)F)(C(F)(F)F)O)N

Specifications

Melting Point 147°C to 152°C
Quantity 5 g
Beilstein 1473737
Formula Weight 259.15
Percent Purity 96%
Chemical Name or Material 2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10189710 Certificate TypeCertificate of Analysis Date13/10/2023 Catalog NumberB24048.03,B24048.14,B24048.06
Lot Number10157793 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24048.03,B24048.14,B24048.06
Lot NumberF09N33 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24048.03,B24048.14,B24048.06
Lot NumberE22R051 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24048.03,B24048.14,B24048.06
Lot Number10139191 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24048.03,B24048.14,B24048.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.