missing translation for 'onlineSavingsMsg'
Learn More
2,4-Dichlorobenzoic acid, 98%, Thermo Scientific Chemicals Product Code.: 10626562

2,4-Dichlorobenzoic acid, 98%, Thermo Scientific Chemicals

Product Code. 10626562
250 g, Plastic bottle
Click to view available options
Quantity:
5 g
100 g
250 g
1 kg
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10626562

Brand: Thermo Scientific Acros 160902500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-84-0
Molecular Formula C7H4Cl2O2
Molecular Weight (g/mol) 191.01
MDL Number MFCD00002414
InChI Key ATCRIUVQKHMXSH-UHFFFAOYSA-N
Synonym benzoic acid, 2,4-dichloro, 2,4-dichlorobenzoesyre, 2,4 dichlorobenzoic acid, 2,4-dichloro-benzoic acid, 2,4-dichlorobenzoicacid, ccris 9472, furosemide impurity e, pubchem21313, acmc-209kqh, wln: qvr bg dgShow MoreShow Less
PubChem CID 5787
ChEBI CHEBI:30748
IUPAC Name 2,4-dichlorobenzoic acid
SMILES C1=CC(=C(C=C1Cl)Cl)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 160.0°C to 164.0°C
Color White
Boiling Point >200.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula Cl2C6H3CO2H
Quantity 250 g
Beilstein 09, 342
Solubility Information Solubility in water: 0.36g/L (15°C). Other solubilities: soluble in alcohol,ether and acetone,insoluble in heptane
Formula Weight 191.01
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2, 4-Dichlorobenzoic acid, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0442746 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number160901000,160900010,160902500,160900050
Lot NumberA0424447 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number160900010,160901000,160902500,160900050
Lot NumberA0434343 Certificate TypeCertificate of Analysis Date15/03/2023 Catalog Number160900010,160901000,160902500,160900050
Lot NumberA0409668 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number160900010,160901000,160902500,160900050
Lot NumberA0389134 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number160900010,160901000,160902500,160900050
compliance-icons
Product Identifier
  • 2,4-Dichlorobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,4-Dichlorobenzoic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.