Learn More
2-(4-Methylperhydro-1,4-diazepin-1-yl)benzylamine, 90%, Thermo Scientific™
95.20 EUR - 114.80 EUR
Chemical Identifiers
CAS | 915707-56-1 |
---|---|
Molecular Formula | C13H21N3 |
Molecular Weight (g/mol) | 219.332 |
MDL Number | MFCD09065001 |
InChI Key | YLJBTEMLBKZLIT-UHFFFAOYSA-N |
Synonym | 2-4-methylperhydro-1,4-diazepin-1-yl benzylamine, 1-2-4-methyl-1,4-diazepan-1-yl phenyl methanamine, 2-4-methylhomopiperazin-1-yl benzylamine, 2-4-methyl-1,4-diazepan-1-yl phenyl methanamine, 2-4-methyl-1,4-diazaperhydroepinyl phenyl methylamine |
PubChem CID | 24229619 |
IUPAC Name | [2-(4-methyl-1,4-diazepan-1-yl)phenyl]methanamine |
SMILES | CN1CCCN(CC1)C2=CC=CC=C2CN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
10747115
|
Thermo Scientific Maybridge
CC60413CB |
250 mg | Amber Glass Bottle | N/A | N/A | N/A | |||||
10581894
|
Thermo Scientific Maybridge
CC60413DA |
1 g | Amber Glass Bottle | N/A | N/A | N/A | |||||
Description
Chemical Identifiers
915707-56-1 | |
219.332 | |
YLJBTEMLBKZLIT-UHFFFAOYSA-N | |
24229619 | |
CN1CCCN(CC1)C2=CC=CC=C2CN |
C13H21N3 | |
MFCD09065001 | |
2-4-methylperhydro-1,4-diazepin-1-yl benzylamine, 1-2-4-methyl-1,4-diazepan-1-yl phenyl methanamine, 2-4-methylhomopiperazin-1-yl benzylamine, 2-4-methyl-1,4-diazepan-1-yl phenyl methanamine, 2-4-methyl-1,4-diazaperhydroepinyl phenyl methylamine | |
[2-(4-methyl-1,4-diazepan-1-yl)phenyl]methanamine |
Specifications
915707-56-1 | |
100.0 | |
C13H21N3 | |
2-4-methylperhydro-1,4-diazepin-1-yl benzylamine, 1-2-4-methyl-1,4-diazepan-1-yl phenyl methanamine, 2-4-methylhomopiperazin-1-yl benzylamine, 2-4-methyl-1,4-diazepan-1-yl phenyl methanamine, 2-4-methyl-1,4-diazaperhydroepinyl phenyl methylamine | |
CN1CCCN(CC1)C2=CC=CC=C2CN | |
219.332 | |
219.33 | |
2-(4-Methylperhydro-1, 4-diazepin-1-yl)benzylamine |
89.5 | |
90% | |
MFCD09065001 | |
YLJBTEMLBKZLIT-UHFFFAOYSA-N | |
[2-(4-methyl-1,4-diazepan-1-yl)phenyl]methanamine | |
24229619 | |
90% |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
Your input is important to us. Please complete this form to provide feedback related to the content on this product.