missing translation for 'onlineSavingsMsg'
Learn More
2,5-Dichloro-3-nitrobenzoic acid, 98+%, Thermo Scientific™ Product Code.: 11304564

2,5-Dichloro-3-nitrobenzoic acid, 98+%, Thermo Scientific™

Product Code. 11304564
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11304564

Brand: Thermo Scientific Alfa Aesar L01332.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-86-8
Molecular Formula C7H3Cl2NO4
Molecular Weight (g/mol) 236.004
MDL Number MFCD00014691
InChI Key AUAXYMOBWXOEQD-UHFFFAOYSA-N
Synonym 2,5-dichloro-3-nitrobenzoic acid, dinoben, benzoic acid, 2,5-dichloro-3-nitro, caswell no. 312, 3-nitro-2,5-dichlorobenzoic acid, unii-5rxk12d272, kyselina 2,5-dichlor-3-nitrobenzoova, epa pesticide chemical code 028101, kyselina 2,5-dichlor-3-nitrobenzoova czech, 2,5-dichloro-3-nitro-benzoic acidShow MoreShow Less
PubChem CID 6951
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])Cl)C(=O)O)Cl

Specifications

Melting Point 218°C to 222°C
Quantity 1 g
Beilstein 1976119
Formula Weight 236.01
Percent Purity ≥98%
Chemical Name or Material 2,5-Dichloro-3-nitrobenzoic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberUSLF002786 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot NumberLF002785 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,5-Dichloro-3-nitrobenzoic acid, 98+%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.