missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dichlorophenyl isocyanate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Specifications
Specifications
CAS | 5392-82-5 |
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Melting Point | 27°C to 29°C |
Color | Colorless to Yellow or White to Yellow |
Boiling Point | 120°C to 125°C (20.0 mmHg) |
Flash Point | 113°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass Bottle |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
335978 | Certificate of Analysis | 05/10/2020 |
Safety and Handling


Product Identifier
- 2,5-Dichlorophenyl isocyanate
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H301-Toxic if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2,5-Dichlorophenyl isocyanate, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction