missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-1-(pyridin-4-ylmethyl)-1H-pyrrole-3-carboxylic acid, 97%, Thermo Scientific™

86.90 EUR - 275.00 EUR

Chemical Identifiers

CAS 306936-15-2
Molecular Formula C13H14N2O2
Molecular Weight (g/mol) 230.267
MDL Number MFCD01567218
InChI Key IHQUENCAKSKBOG-UHFFFAOYSA-N
Synonym 2,5-dimethyl-1-pyridin-4-ylmethyl-1h-pyrrole-3-carboxylic acid, 2,5-dimethyl-1-pyridin-4-ylmethyl pyrrole-3-carboxylic acid, maybridge1_008545, 2,5-dimethyl-1-4-pyridylmethyl pyrrole-3-carboxylic acid, 1h-pyrrole-3-carboxylicacid, 2,5-dimethyl-1-4-pyridinylmethyl, 2,5-dimethyl-1-pyridin-4-yl methyl-1h-pyrrole-3-carboxylic acidShow MoreShow Less
PubChem CID 2736289
IUPAC Name 2,5-dimethyl-1-(pyridin-4-ylmethyl)pyrrole-3-carboxylic acid
SMILES CC1=CC(=C(N1CC2=CC=NC=C2)C)C(=O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10691503
SDS View Documents
Thermo Scientific Maybridge
SP00292DA
1 g
86.90 EUR
1g
Please to purchase this item. Need a web account? Register with us today!
10260305
SDS View Documents
Thermo Scientific Maybridge
SP00292DE
5 g
275.00 EUR
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications
Show More
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,5-Dimethyl-1-(pyridin-4-ylmethyl)-1H-pyrrole-3-carboxylic acid, 97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.