missing translation for 'onlineSavingsMsg'
Learn More
2,6-Dimethyl-4-heptanone, >90% (sum of 2,6-Dimethyl-4-heptanone & 4,6-Dimethyl-2-heptanone), Thermo Scientific Chemicals Product Code.: 11332857

2,6-Dimethyl-4-heptanone, >90% (sum of 2,6-Dimethyl-4-heptanone & 4,6-Dimethyl-2-heptanone), Thermo Scientific Chemicals

Product Code. 11332857
500 mL
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
This item is not returnable. View return policy

Product Code. 11332857

Brand: Thermo Scientific Alfa Aesar L16015.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2,6-Dimethyl-4-heptanone is used as a coating solvent. It is an active component of mint oil. It acts as a dispersant for organosol type resins. It is involved in the antigerminative treatment of bulbs and tubers. Further, it is used as a solvent for nitrocellulose. In addition to this, it acts as an intermediate in the preparation of inhibitors, active pharmaceutical ingredients and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethyl-4-heptanone is used as a coating solvent. It is an active component of mint oil. It acts as a dispersant for organosol type resins. It is involved in the antigerminative treatment of bulbs and tubers. Further, it is used as a solvent for nitrocellulose. In addition to this, it acts as an intermediate in the preparation of inhibitors, active pharmaceutical ingredients and dyes.

Solubility
Miscible with ethanol, ether, carbon tetrachloride, chloroform, benzene and most organic liquids.

Notes
Incompatible with strong bases, strong oxidizing agents, strong acids and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-83-8
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.242
MDL Number MFCD00008940
InChI Key PTTPXKJBFFKCEK-UHFFFAOYSA-N
Synonym 2,6-dimethyl-4-heptanone, diisobutyl ketone, isovalerone, isobutyl ketone, valerone, diisobutylketone, diisobutilchetone, s-diisopropylacetone, di-isobutylcetone, dibkShow MoreShow Less
PubChem CID 7958
IUPAC Name 2,6-dimethylheptan-4-one
SMILES CC(C)CC(=O)CC(C)C

Specifications

Melting Point -42°C
Density 0.809
Boiling Point 163°C to 173°C
Flash Point 47°C (117°F)
Odor Unpleasant
Refractive Index 1.414
Quantity 500 mL
UN Number UN1224
Beilstein 1743163
Solubility Information Miscible with ethanol,ether,carbon tetrachloride,chloroform,benzene and most organic liquids.
Formula Weight 142.24
Percent Purity >90%
Chemical Name or Material 2,6-Dimethyl-4-heptanone, (sum of 2,6-Dimethyl-4-heptanone & 4,6-Dimethyl-2-heptanone)
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250828 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL16015.AE,L16015.AP,L16015.0F
Lot NumberS27K022 Certificate TypeCertificate of Analysis Date23/07/2024 Catalog NumberL16015.AE,L16015.AP,L16015.0F
Lot NumberS05K069 Certificate TypeCertificate of Analysis Date17/06/2024 Catalog NumberL16015.AE,L16015.AP,L16015.0F
Lot NumberP18K019 Certificate TypeCertificate of Analysis Date12/06/2024 Catalog NumberL16015.AE,L16015.AP,L16015.0F
Lot NumberP11K045 Certificate TypeCertificate of Analysis Date18/03/2024 Catalog NumberL16015.AE,L16015.AP,L16015.0F
compliance-icons compliance-icons
Product Identifier
  • 2,6-Dimethyl-4-heptanone, (sum of 2,6-Dimethyl-4-heptanone & 4,6-Dimethyl-2-heptanone)
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contains : Diisobutyl ketone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,6-Dimethyl-4-heptanone, >90% (sum of 2,6-Dimethyl-4-heptanone & 4,6-Dimethyl-2-heptanone), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.