missing translation for 'onlineSavingsMsg'
Learn More

2-Allyl-N-Fmoc-L-glycine, 95%, Thermo Scientific Chemicals

62.51 EUR - 699.48 EUR

Chemical Identifiers

CAS 146549-21-5
Molecular Formula C20H19NO4
Molecular Weight (g/mol) 337.375
MDL Number MFCD01311749
InChI Key YVBLQCANYSFEBN-SFHVURJKSA-N
Synonym fmoc-l-allylglycine, fmoc-gly allyl-oh, fmoc-l-allyglycine, s-2-9h-fluoren-9-yl methoxy carbonyl amino pent-4-enoic acid, fmoc-allyl-gly-oh, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino pent-4-enoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino pent-4-enoic acid, 4-pentenoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino-, 2s, s-n-fmoc-allyl-glycine, s-n-fmoc-allylglycine
PubChem CID 2734457
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pent-4-enoic acid
SMILES C=CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15488727
SDS View Documents
Thermo Scientific Alfa Aesar
H52177.MD
250 mg N/A N/A N/A
15468727
SDS View Documents
Thermo Scientific Alfa Aesar
H52177.03
1 g
190.95 EUR
1g
Please to purchase this item. Need a web account? Register with us today!
15478727
SDS View Documents
Thermo Scientific Alfa Aesar
H52177.06
5 g
699.48 EUR
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2-Allyl-N-Fmoc-L-glycine is potentially useful for solid phase peptide synthesis techniques. 2-Allyl-N-Fmoc-L-glycine compound could be useful as an unusual amino acid analog to aid in the deconvolution of protein structure and function. Reagent in the synthesis of a Dicarba-Analog of Octreotide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Allyl-N-Fmoc-L-glycine is potentially useful for solid phase peptide synthesis techniques. 2-Allyl-N-Fmoc-L-glycine compound could be useful as an unusual amino acid analog to aid in the deconvolution of protein structure and function. Reagent in the synthesis of a Dicarba-Analog of Octreotide.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Allyl-N-Fmoc-L-glycine, 95%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.