missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-3-bromobenzoic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 10 g |
Formula Weight | 216.03 |
Percent Purity | 97% |
Chemical Name or Material | 2-Amino-3-bromobenzoic acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2-Amino-3-bromobenzoic acid, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction