missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chloro-3-nitropyridine, 95%, Thermo Scientific Chemicals

39.55 EUR - 378.89 EUR

Chemical Identifiers

CAS 6980-08-1
Molecular Formula C5H4ClN3O2
Molecular Weight (g/mol) 173.56
MDL Number MFCD04116097
InChI Key DIRINUVNYFAWQF-UHFFFAOYSA-N
Synonym 2-amino-4-chloro-3-nitropyridine, 2-amino-3-nitro-4-chloropyridine, 4-chloro-3-nitro-2-pyridinamine, 4-chloro-3-nitro-pyridin-2-ylamine, 2-pyridinamine, 4-chloro-3-nitro, 4-chloro-3-nitro-pyridin-2-amine, pubchem17739, ksc352m6b, 3-nitro-4-chloropyridine-2-amine, 4-chloro-2-amino-3-nitropyridine
PubChem CID 4071694
IUPAC Name 4-chloro-3-nitropyridin-2-amine
SMILES C1=CN=C(C(=C1Cl)[N+](=O)[O-])N
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15431989
SDS View Documents
Thermo Scientific Alfa Aesar
H64678.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15411989
SDS View Documents
Thermo Scientific Alfa Aesar
H64678.03
1 g
104.52 EUR
1g
Please to purchase this item. Need a web account? Register with us today!
15421989
SDS View Documents
Thermo Scientific Alfa Aesar
H64678.06
5 g
378.89 EUR
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2-Amino-4-chloro-3-nitropyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4-chloro-3-nitropyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in dimethyl formamide, dimethyl sulfoxide, hot ethanol, ethyl acetate and hot methanol.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Amino-4-chloro-3-nitropyridine, 95%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.