missing translation for 'onlineSavingsMsg'
Learn More
2-Amino-5-bromo-3-methylpyridine, 97%, Thermo Scientific Chemicals Product Code.: 11458206

2-Amino-5-bromo-3-methylpyridine, 97%, Thermo Scientific Chemicals

Product Code. 11458206
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11458206

Brand: Thermo Scientific Alfa Aesar A15318.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3430-21-5
MDL Number MFCD00068232
Synonym 2-amino-5-bromo-3-methylpyridine, 2-amino-5-bromo-3-picoline, 2-amino-3-methyl-5-bromopyridine, 5-bromo-3-methyl-pyridin-2-ylamine, 2-pyridinamine, 5-bromo-3-methyl, 5-bromo-3-methyl-2-pyridinamine, 2-amina-5-bromo-3-picoline, 5-bromo-3-methyl-2-pyridylamine, 5-bromo-3-methyl-pyridin-2-amine, 5-bromo-3-methylpyridin-2-ylamine
PubChem CID 137934

Specifications

Melting Point 90°C to 94°C
Odor Unpleasant
Molecular Formula C6H7BrN2
Quantity 1 g
Beilstein 386426
Sensitivity Hygroscopic
InChI Key KBLGGRWUEVCNPY-UHFFFAOYSA-N
SMILES CC1=CC(=CN=C1N)Br
IUPAC Name 5-bromo-3-methylpyridin-2-amine
Molecular Weight (g/mol) 187.04
Formula Weight 187.04
Percent Purity 97%
Chemical Name or Material 2-Amino-5-bromo-3-methylpyridine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Amino-5-bromo-3-methylpyridine, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.