missing translation for 'onlineSavingsMsg'
Learn More
2-Amino-6-chlorobenzoic acid, 97+%, Thermo Scientific Chemicals Product Code.: 11319689

2-Amino-6-chlorobenzoic acid, 97+%, Thermo Scientific Chemicals

Product Code. 11319689
10 g
Click to view available options
Quantity:
5 g
10 g
25 g
This item is not returnable. View return policy

Product Code. 11319689

Brand: Thermo Scientific Alfa Aesar A10890.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Amino-6-chlorobenzoic acid is used as intermediate for medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-6-chlorobenzoic acid is used as intermediate for medicine.

Solubility
Very soluble in water. Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2148-56-3
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00051530
InChI Key SZCPTRGBOVXVCA-UHFFFAOYSA-N
Synonym 6-chloroanthranilic acid, benzoic acid, 2-amino-6-chloro, 2-amino-6-chloro-benzoic acid, 6-chloranthranilsaure, pubchem9876, acmc-1cngq, 6-chloro anthranilic acid, 2-carboxy-3-chloroaniline, 2amino-6-chlorobenzoic acid, ksc201s6bShow MoreShow Less
PubChem CID 75071
IUPAC Name 2-amino-6-chlorobenzoic acid
SMILES C1=CC(=C(C(=C1)Cl)C(=O)O)N

Specifications

Melting Point 149°C to 152°C
Quantity 10 g
Beilstein 2804041
Solubility Information Very soluble in water. Soluble in methanol.
Formula Weight 171.58
Percent Purity 98%
Chemical Name or Material 2-Amino-6-chlorobenzoic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA26Q01A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10890.09,A10890.06,A10890.14
Lot Number10227401 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10890.09,A10890.06,A10890.14
Lot Number10231579 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10890.09,A10890.06,A10890.14
Lot Number10129317 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10890.09,A10890.06,A10890.14
Lot Number10225354 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10890.09,A10890.06,A10890.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Amino-6-chlorobenzoic acid, 97+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.