Learn More
2-Aminoisobutyric acid, 99%, Thermo Scientific Chemicals
46.50 EUR - 218.00 EUR
Chemical Identifiers
CAS | 62-57-7 |
---|---|
Molecular Formula | C4H9NO2 |
Molecular Weight (g/mol) | 103.121 |
MDL Number | MFCD00008049 |
InChI Key | FUOOLUPWFVMBKG-UHFFFAOYSA-N |
Synonym | 2-aminoisobutyric acid, 2-methylalanine, alanine, 2-methyl, alpha-aminoisobutyric acid, 2-amino-2-methylpropionic acid, h-aib-oh, alpha-methylalanine, alpha-aminoisobutanoic acid, a-aminoisobutyric acid, 2-amino isobutyric acid |
PubChem CID | 6119 |
ChEBI | CHEBI:27971 |
IUPAC Name | 2-amino-2-methylpropanoic acid |
SMILES | CC(C)(C(=O)O)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11389679
|
Thermo Scientific Alfa Aesar
A13021.14 |
25 g |
46.50 EUR
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11399679
|
Thermo Scientific Alfa Aesar
A13021.22 |
100 g |
119.00 EUR
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11489933
|
Thermo Scientific Alfa Aesar
A13021.30 |
250 g |
218.00 EUR
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Aminoisobutyric acid is a non- protein amino acid present in some antibiotics. It acts as a strong helix inducer in peptides. It is non-metabolisable used in bioassays.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Aminoisobutyric acid is a non- protein amino acid present in some antibiotics. It acts as a strong helix inducer in peptides. It is non-metabolisable used in bioassays.
Solubility
Soluble in water. Insoluble in alcohol and ether.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
62-57-7 | |
103.121 | |
FUOOLUPWFVMBKG-UHFFFAOYSA-N | |
6119 | |
2-amino-2-methylpropanoic acid |
C4H9NO2 | |
MFCD00008049 | |
2-aminoisobutyric acid, 2-methylalanine, alanine, 2-methyl, alpha-aminoisobutyric acid, 2-amino-2-methylpropionic acid, h-aib-oh, alpha-methylalanine, alpha-aminoisobutanoic acid, a-aminoisobutyric acid, 2-amino isobutyric acid | |
CHEBI:27971 | |
CC(C)(C(=O)O)N |
Specifications
62-57-7 | |
C4H9NO2 | |
506496 | |
2-aminoisobutyric acid, 2-methylalanine, alanine, 2-methyl, alpha-aminoisobutyric acid, 2-amino-2-methylpropionic acid, h-aib-oh, alpha-methylalanine, alpha-aminoisobutanoic acid, a-aminoisobutyric acid, 2-amino isobutyric acid | |
FUOOLUPWFVMBKG-UHFFFAOYSA-N | |
2-amino-2-methylpropanoic acid | |
6119 | |
103.12 | |
2-Aminoisobutyric acid |
>300°C | |
MFCD00008049 | |
14,445 | |
Soluble in water. Insoluble in alcohol and ether. | |
CC(C)(C(=O)O)N | |
103.121 | |
CHEBI:27971 | |
99% |
Safety and Handling
missing translation for 'einecsNumber' : 200-544-0
missing translation for 'rtecsNumber' : AY7000000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only