Learn More
2-Bromo-4-(2-thienyl)thiazole, 97%, Thermo Scientific™ Product Code.: 15434818
2-Bromo-4-(2-thienyl)thiazole, 97%, Thermo Scientific™
Product Code. 15434818
2 g
Click to view available options
Quantity:
2 g
10 g
Product Code. 15434818
Brand: Thermo Scientific Alfa Aesar H58575.04
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1125409-90-6
Molecular Formula C7H4BrNS2
Molecular Weight (g/mol) 246.14
MDL Number MFCD11220135
InChI Key XRXGEQOSIYEEKD-UHFFFAOYSA-N
Synonym 2-bromo-4-2-thienyl thiazole, 2-bromo-4-thiophen-2-yl-1,3-thiazole
PubChem CID 38996210
IUPAC Name 2-bromo-4-thiophen-2-yl-1,3-thiazole
SMILES C1=CSC(=C1)C2=CSC(=N2)Br
Specifications
Quantity 2 g
Beilstein 21223492
Formula Weight 246.14
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-(2-thienyl)thiazole
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR09C034 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction
Product Title
2-Bromo-4-(2-thienyl)thiazole, 97%, Thermo Scientific™ >