missing translation for 'onlineSavingsMsg'
Learn More
2-Chloro-3-nitro-5-(trifluoromethyl)pyridine, 95%, Thermo Scientific Chemicals Product Code.: 15421589

2-Chloro-3-nitro-5-(trifluoromethyl)pyridine, 95%, Thermo Scientific Chemicals

Product Code. 15421589
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 15421589

Brand: Thermo Scientific Alfa Aesar H64394.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72587-15-6
Molecular Formula C6H2ClF3N2O2
Molecular Weight (g/mol) 226.539
MDL Number MFCD06255171
InChI Key DRPIKFKCAJGTJF-UHFFFAOYSA-N
Synonym 2-chloro-3-nitro-5-trifluoromethyl pyridine, 2-chloro-5-trifluoromethyl-3-nitropyridine, pyridine, 2-chloro-3-nitro-5-trifluoromethyl, 5-trifluoromethyl-3-nitro-2-chloropyridine, pubchem17529, acmc-209onn, abbypharma ap-11-5568, 2-chloro-3-nitro-5-trifluoromethyl-pyridine, 2-chloro3-nitro-5-trifluoromethyl pyridineShow MoreShow Less
PubChem CID 4736950
IUPAC Name 2-chloro-3-nitro-5-(trifluoromethyl)pyridine
SMILES C1=C(C=NC(=C1[N+](=O)[O-])Cl)C(F)(F)F

Specifications

Color Colorless to Yellow
Quantity 5 g
Beilstein 7586778
Formula Weight 226.54
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Chloro-3-nitro-5-(trifluoromethyl)pyridine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number81302936 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH64394.06,H64394.03,H64394.14
Lot NumberQ27G038 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberH64394.06,H64394.03,H64394.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Chloro-3-nitro-5-(trifluoromethyl)pyridine, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.