missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(trifluoromethyl)nicotinic acid, 97%, Thermo Scientific™

Product Code. 11372389
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590371-81-6
Molecular Formula C7H3ClF3NO2
Molecular Weight (g/mol) 225.55
MDL Number MFCD08458125
InChI Key FZIJHEOXLKCIRZ-UHFFFAOYSA-N
Synonym 2-chloro-4-trifluoromethyl nicotinic acid, 2-chloro-4-trifluoromethyl pyridine-3-carboxylic acid, 3-carboxy-2-chloro-4-trifluoromethyl pyridine, acmc-1az7h, ksc269c2b, 3-pyridinecarboxylicacid, 2-chloro-4-trifluoromethyl, 3-pyridinecarboxylic acid, 2-chloro-4-trifluoromethyl, 2-chloro-4-trifluoromethyl-3-pyridinecarboxylic acid
PubChem CID 11770138
IUPAC Name 2-chloro-4-(trifluoromethyl)pyridine-3-carboxylic acid
SMILES OC(=O)C1=C(C=CN=C1Cl)C(F)(F)F

Specifications

Melting Point 158°C to 159°C
Quantity 1 g
Formula Weight 225.56
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-(trifluoromethyl)nicotinic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Chloro-4-(trifluoromethyl)nicotinic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.