missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-5-(methoxycarbonyl)benzeneboronic acid, 98%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Boronic acids are often used in metal-catalyzed cross-coupling for C-C, C-N bond formation, and C-H functionalization. The widely utilized Suzuki-Miyaura coupling allows for the coupling of boronic acids with carbon halides and surrogates. Other known chemistry may includes 1,4-addition to activated alkenes, 1,2-addition to carbonyls and imines, addition to anhydrides, 1,4-addition to a,ß-unsaturated ketones(4), and 1,4-addition to esters and amides.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Boronic acids are often used in metal-catalyzed cross-coupling for C-C, C-N bond formation, and C-H functionalization. The widely utilized Suzuki-Miyaura coupling allows for the coupling of boronic acids with carbon halides and surrogates. Other known chemistry may includes 1,4-addition to activated alkenes, 1,2-addition to carbonyls and imines, addition to anhydrides, 1,4-addition to a,ß-unsaturated ketones(4), and 1,4-addition to esters and amides.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Specifications
Specifications
Melting Point | 230°C to 232°C |
Quantity | 250 mg |
Formula Weight | 214.41 |
Percent Purity | 98% |
Chemical Name or Material | 2-Chloro-5-(methoxycarbonyl)benzeneboronic acid |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
B-BC080202 | Certificate of Analysis | 07/12/2021 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2-Chloro-5-(methoxycarbonyl)benzeneboronic acid, 98%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction