missing translation for 'onlineSavingsMsg'
Learn More
2-Chlorophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals Product Code.: 11428106

2-Chlorophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Product Code. 11428106
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11428106

Brand: Thermo Scientific Alfa Aesar A14136.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Chlorophenylhydrazine hydrochloride is used to study intrinsic resistance of Mycobacterium smegmatis to fluoroquinolones, used for treatment of Mycobacterium tuberculosis. It may be used in pyrazoline synthesis. It can be used to produce N-azepan-2-ylidene-N'-(2-chloro-phenyl)-hydrazine with 7-methoxy-3,4,5,6-tetrahydro-2H-azepine at heating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorophenylhydrazine hydrochloride is used to study intrinsic resistance of Mycobacterium smegmatis to fluoroquinolones, used for treatment of Mycobacterium tuberculosis. It may be used in pyrazoline synthesis. It can be used to produce N-azepan-2-ylidene-N′-(2-chloro-phenyl)-hydrazine with 7-methoxy-3,4,5,6-tetrahydro-2H-azepine at heating.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41052-75-9
Molecular Formula C6H8Cl2N2
Molecular Weight (g/mol) 179.04
MDL Number MFCD00012928
InChI Key ADODRSVGNHNKAT-UHFFFAOYSA-N
Synonym 2-chlorophenylhydrazine hydrochloride, 2-chlorophenyl hydrazine hydrochloride, o-chlorophenylhydrazine hydrochloride, 1-2-chlorophenyl hydrazine hydrochloride, o-chlorophenylhydrazine hcl, 2-chloro phenyl hydrazine hydrochloride, 2-chlorophenylhydrazine, chloride, hydrazine, 2-chlorophenyl-, monohydrochloride, 2-chlorophenylhydrazinehydrochloride
PubChem CID 443287
ChEBI CHEBI:1046
IUPAC Name (2-chlorophenyl)hydrazine;hydrochloride
SMILES [H+].[Cl-].NNC1=CC=CC=C1Cl

Specifications

Melting Point ∼201°C (decomposition)
Quantity 25 g
Beilstein 3699381
Solubility Information Soluble in water.
Formula Weight 179.05
Percent Purity 97%
Chemical Name or Material 2-Chlorophenylhydrazine hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10212930 Certificate TypeCertificate of Analysis Date20/07/2023 Catalog NumberA14136.14,A14136,A14136.06,A14136.22
Lot Number10212930 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14136.14,A14136.06,A14136.22
Lot Number10193620 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14136.14,A14136.06,A14136.22
Lot Number10194113 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14136.14,A14136.06,A14136.22
Lot NumberD2057B Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14136.14,A14136.06,A14136.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Chlorophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.