missing translation for 'onlineSavingsMsg'
Learn More
2-Chloropyridine-4-boronic acid pinacol ester, 97%, Thermo Scientific™ Product Code.: 15259906

2-Chloropyridine-4-boronic acid pinacol ester, 97%, Thermo Scientific™

Product Code. 15259906
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15259906

Brand: Thermo Scientific Acros 454220050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 458532-84-8
Molecular Formula C11H15BClNO2
Molecular Weight (g/mol) 239.51
InChI Key UUEQDBHKMOFLDP-UHFFFAOYSA-N
Synonym 2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-chloropyridine-4-boronic acid pinacol ester, 2-chloro-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-chloropyridin-4-ylboronic acid pinacol ester, 2-chloro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2-chloropyridine-4-boronic acid, pinacol ester, 2-chloro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine, pyridine, 2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pubchem16684
PubChem CID 5103408
IUPAC Name 2-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=NC=C2)Cl

Specifications

Melting Point 63.5°C to 66.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 239.51
Percent Purity 97%
Chemical Name or Material 2-Chloropyridine-4-boronic acid pinacol ester
compliance-icons
Product Identifier
  • 2-Chloropyridine-4-boronic acid pinacol ester, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Chloropyridine-4-boronic acid pinacol ester, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.