missing translation for 'onlineSavingsMsg'
Learn More
2-Chloroquinoline-3-carboxaldehyde, 98%, Thermo Scientific Chemicals Product Code.: 11456240

2-Chloroquinoline-3-carboxaldehyde, 98%, Thermo Scientific Chemicals

Product Code. 11456240
1 g
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy

Product Code. 11456240

Brand: Thermo Scientific Alfa Aesar H27133.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73568-25-9
Molecular Formula C10H6ClNO
Molecular Weight (g/mol) 191.614
MDL Number MFCD00130079
InChI Key SDKQWXCBSNMYBN-UHFFFAOYSA-N
Synonym 2-chloro-3-quinolinecarboxaldehyde, 2-chloro-3-quinoline carboxaldehyde, 2-chloro-quinoline-3-carbaldehyde, 2-chloroquinoline-3-carboxaldehyde, 3-quinolinecarboxaldehyde, 2-chloro, 2-chloro-3-quinolinecarbaldehyde, acmc-209or9, 2-chloro-3-formylquinoline, ksc494c9n, 2-chloroquinolin-3-carbaldehyde
PubChem CID 690958
IUPAC Name 2-chloroquinoline-3-carbaldehyde
SMILES C1=CC=C2C(=C1)C=C(C(=N2)Cl)C=O

Specifications

Melting Point 148°C to 150°C
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 191.62
Percent Purity 98%
Chemical Name or Material 2-Chloroquinoline-3-carboxaldehyde

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10244653 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog NumberH27133.09,H27133.03
Lot Number10135898 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27133.09,H27133.03
Lot Number10167853 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27133.09,H27133.03
Lot Number10228271 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27133.09,H27133.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Chloroquinoline-3-carboxaldehyde, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.