missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals 2'-Deoxyguanosine-5'-monophosphate disodium salt hydrate, 98% Product Code.: 15480417

Thermo Scientific Chemicals 2'-Deoxyguanosine-5'-monophosphate disodium salt hydrate, 98%

Product Code. 15480417
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 15480417

Brand: Thermo Scientific Chemicals J65186.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2'-Deoxyguanosine-5'-monophosphate Disodium Salt Hydrate is a reactant involved in analysis of self-assembling in solution and nucleation/growth of G-qudruplexes, Nucleophilic trapping, Reductive alkylation. It is a nucleotide precursor used in DNA synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Deoxyguanosine-5′-monophosphate Disodium Salt Hydrate is a reactant involved in analysis of self-assembling in solution and nucleation/growth of G-qudruplexes, Nucleophilic trapping, Reductive alkylation. It is a nucleotide precursor used in DNA synthesis.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146877-98-7
Molecular Formula C10H12N5Na2O7P
Molecular Weight (g/mol) 391.19
MDL Number MFCD00080331,MFCD00150758
InChI Key CTPAMSRBXKGZCJ-UHFFFAOYNA-L
Synonym 2'-Deoxy-5'-guanylic acid disodium salt; 5'-dGMP disodium salt
PubChem CID 123133822
IUPAC Name [(2R,3S,5R)-5-(2-amino-6-oxo-3H-purin-9-yl)-3-hydroxyoxolan-2-yl]methyl dihydrogen phosphate;sodium
SMILES [Na+].[Na+].NC1=NC(=O)C2=C(N1)N(C=N2)C1CC(O)C(COP([O-])([O-])=O)O1

Specifications

Melting Point >245°C (decomposition)
Color White
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 391.18 (Anhydrous)
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2'-Deoxyguanosine-5'-monophosphate disodium salt hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX05J010 Certificate TypeCertificate of Analysis Date11/10/2023 Catalog NumberJ65186.06,J65186.03,J65186.14
Lot NumberN28J022 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberJ65186.06
Lot NumberZ09F054 Certificate TypeCertificate of Analysis Date03/02/2023 Catalog NumberJ65186.06,J65186.03,J65186.14
Lot NumberY29I032 Certificate TypeCertificate of Analysis Date06/01/2023 Catalog NumberJ65186.06,J65186.03,J65186.14
Lot NumberP16B006 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ65186.06,J65186.03,J65186.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals 2'-Deoxyguanosine-5'-monophosphate disodium salt hydrate, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.