missing translation for 'onlineSavingsMsg'
Learn More
2-Fluoropyridine-3-boronic acid, 97%, Thermo Scientific Chemicals Product Code.: 11384546

2-Fluoropyridine-3-boronic acid, 97%, Thermo Scientific Chemicals

Product Code. 11384546
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11384546

Brand: Thermo Scientific Alfa Aesar L20108.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 174669-73-9
Molecular Formula C5H5BFNO2
Molecular Weight (g/mol) 140.91
MDL Number MFCD03411557
InChI Key YUHZIUAREWNXJT-UHFFFAOYSA-N
Synonym 2-fluoropyridine-3-boronic acid, 2-fluoro-3-pyridylboronic acid, 2-fluoropyridin-3-yl boronic acid, 2-fluoro-3-pyridineboronic acid, 2-fluoropyridin-3-yl-3-boronic acid, 2-fluoro-3-pyridilboronic acid, 2-fluoropryridine-3-boronic acid, boronic acid, 2-fluoro-3-pyridinyl, 2-fluoro-3-pyridyl boronic acid, 2-fluoropyridine-3-boronicacid
PubChem CID 2783270
IUPAC Name (2-fluoropyridin-3-yl)boronic acid
SMILES OB(O)C1=CC=CN=C1F

Specifications

Melting Point 172°C
Quantity 1 g
Beilstein 9119935
Formula Weight 140.91
Percent Purity 97%
Chemical Name or Material 2-Fluoropyridine-3-boronic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10111353 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL20108.03,L20108.06
Lot NumberF15R029 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL20108.03,L20108.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Fluoropyridine-3-boronic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.