Learn More
2-Hydroxy-3-nitro-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific™
66.10 EUR - 789.00 EUR
Chemical Identifiers
CAS | 33252-64-1 |
---|---|
Molecular Formula | C6H3F3N2O3 |
Molecular Weight (g/mol) | 208.096 |
MDL Number | MFCD00153193 |
InChI Key | JYXKHKBZLLIWEV-UHFFFAOYSA-N |
PubChem CID | 2775093 |
IUPAC Name | 3-nitro-5-(trifluoromethyl)-1H-pyridin-2-one |
SMILES | C1=C(C(=O)NC=C1C(F)(F)F)[N+](=O)[O-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
15408456
|
Thermo Scientific Alfa Aesar
H63694.MD |
250 mg | N/A | N/A | N/A | |||||
15400529
|
Thermo Scientific Alfa Aesar
H63694.03 |
1 g | N/A | N/A | N/A | |||||
15410529
|
Thermo Scientific Alfa Aesar
H63694.06 |
5 g | N/A | N/A | N/A | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
33252-64-1 | |
208.096 | |
JYXKHKBZLLIWEV-UHFFFAOYSA-N | |
3-nitro-5-(trifluoromethyl)-1H-pyridin-2-one |
C6H3F3N2O3 | |
MFCD00153193 | |
2775093 | |
C1=C(C(=O)NC=C1C(F)(F)F)[N+](=O)[O-] |
Specifications
33252-64-1 | |
White | |
MFCD00153193 | |
2-hydroxy-3-nitro-5-trifluoromethyl pyridine, 3-nitro-5-trifluoromethyl pyridin-2 1h-one, 3-nitro-5-trifluoromethyl pyridin-2-ol, 3-nitro-5-trifluoromethyl-2-pyridinone, 2 1h-pyridinone,3-nitro-5-trifluoromethyl, 5-trifluoromethyl-3-nitropyridin-2-ol, 3-nitro-5-trifluoromethyl-1h-pyridin-2-one, 2 1h-pyridinone, 3-nitro-5-trifluoromethyl, 3-nitro-5-trifluoromethyl hydropyridin-2-one, pubchem9284 | |
C1=C(C(=O)NC=C1C(F)(F)F)[N+](=O)[O-] | |
208.096 | |
208.09 | |
Solid |
189°C to 190°C | |
C6H3F3N2O3 | |
UN2811 | |
JYXKHKBZLLIWEV-UHFFFAOYSA-N | |
3-nitro-5-(trifluoromethyl)-1H-pyridin-2-one | |
2775093 | |
97% | |
2-Hydroxy-3-nitro-5-(trifluoromethyl)pyridine |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.