Learn More
2-Hydroxy-4-(methacryloyloxy)benzophenone, 99%, Thermo Scientific Chemicals
93.67 EUR - 1249.22 EUR
Chemical Identifiers
CAS | 2035-72-5 |
---|---|
Molecular Formula | C17H14O4 |
Molecular Weight (g/mol) | 282.295 |
MDL Number | MFCD00080561 |
InChI Key | IMNBHNRXUAJVQE-UHFFFAOYSA-N |
Synonym | 4-benzoyl-3-hydroxyphenyl methacrylate, 4-methacryloxy-2-hydroxybenzophenone, 2-hydroxy-4-methacryloyloxy benzophenone, methacrylic acid 4-benzoyl-3-hydroxyphenyl ester, 3-hydroxy-4-phenylcarbonyl phenyl 2-methylprop-2-enoate, 2-propenoic acid, 2-methyl-, 4-benzoyl-3-hydroxyphenyl ester, acmc-20amfp, 2-hydroxy-4-methacryloxybenzophenone, 2-hydroxy-4-methacryloyloxybenzophenoneShow More |
PubChem CID | 74856 |
IUPAC Name | (4-benzoyl-3-hydroxyphenyl) 2-methylprop-2-enoate |
SMILES | CC(=C)C(=O)OC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11470597
|
Thermo Scientific Alfa Aesar
B22445.06 |
5 g |
93.67 EUR
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11480597
|
Thermo Scientific Alfa Aesar
B22445.14 |
25 g |
363.81 EUR
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11490597
|
Thermo Scientific Alfa Aesar
B22445.22 |
100 g |
1249.22 EUR
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Hydroxy-4-(methacryloyloxy)benzophenone is used in ophthalmic devices, films, coatings, and fibers, cosmetic preparations and toners.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Hydroxy-4-(methacryloyloxy)benzophenone is used in ophthalmic devices, films, coatings, and fibers, cosmetic preparations and toners.
Solubility
Insoluble in water.
Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
2035-72-5 | |
282.295 | |
IMNBHNRXUAJVQE-UHFFFAOYSA-N | |
74856 | |
CC(=C)C(=O)OC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O |
C17H14O4 | |
MFCD00080561 | |
4-benzoyl-3-hydroxyphenyl methacrylate, 4-methacryloxy-2-hydroxybenzophenone, 2-hydroxy-4-methacryloyloxy benzophenone, methacrylic acid 4-benzoyl-3-hydroxyphenyl ester, 3-hydroxy-4-phenylcarbonyl phenyl 2-methylprop-2-enoate, 2-propenoic acid, 2-methyl-, 4-benzoyl-3-hydroxyphenyl ester, acmc-20amfp, 2-hydroxy-4-methacryloxybenzophenone, 2-hydroxy-4-methacryloyloxybenzophenoneShow More | |
(4-benzoyl-3-hydroxyphenyl) 2-methylprop-2-enoate |
Specifications
2035-72-5 | |
1.18 | |
MFCD00080561 | |
4-benzoyl-3-hydroxyphenyl methacrylate, 4-methacryloxy-2-hydroxybenzophenone, 2-hydroxy-4-methacryloyloxy benzophenone, methacrylic acid 4-benzoyl-3-hydroxyphenyl ester, 3-hydroxy-4-phenylcarbonyl phenyl 2-methylprop-2-enoate, 2-propenoic acid, 2-methyl-, 4-benzoyl-3-hydroxyphenyl ester, acmc-20amfp, 2-hydroxy-4-methacryloxybenzophenone, 2-hydroxy-4-methacryloyloxybenzophenone | |
IMNBHNRXUAJVQE-UHFFFAOYSA-N | |
(4-benzoyl-3-hydroxyphenyl) 2-methylprop-2-enoate | |
74856 | |
99% |
77°C to 79°C | |
C17H14O4 | |
Light sensitive | |
Insoluble in water. | |
CC(=C)C(=O)OC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O | |
282.295 | |
282.3 | |
2-Hydroxy-4-(methacryloyloxy)benzophenone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 218-000-6
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.