missing translation for 'onlineSavingsMsg'
Learn More
2-Hydroxy-6-methylisonicotinic acid, 97%, Thermo Scientific™ Product Code.: 10479454

2-Hydroxy-6-methylisonicotinic acid, 97%, Thermo Scientific™

Product Code. 10479454
5 g
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy

Product Code. 10479454

Brand: Thermo Scientific Maybridge MO00027DE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 86454-13-9
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD02682019
InChI Key NQXYVTIQDNOHFM-UHFFFAOYSA-N
Synonym 2-hydroxy-6-methylisonicotinic acid, 2-hydroxy-6-methylpyridine-4-carboxylic acid, 6-methyl-2-oxo-1,2-dihydropyridine-4-carboxylic acid, 2-hydroxy-6-methyl-isonicotinicacid, 6-hydroxy-2-methylpyridine-4-carboxylic acid, 1,2-dihydro-6-methyl-2-oxopyridine-4-carboxylic acid, 4-pyridinecarboxylic acid, 1,2-dihydro-6-methyl-2-oxo, 4-pyridinecarboxylicacid, 1,2-dihydro-6-methyl-2-oxo, maybridge3_004231, acmc-209q9qShow MoreShow Less
PubChem CID 276660
IUPAC Name 2-methyl-6-oxo-1H-pyridine-4-carboxylic acid
SMILES CC1=CC(=CC(=O)N1)C(O)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 153.14
Percent Purity 97%
Chemical Name or Material 2-Hydroxy-6-methylisonicotinic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Hydroxy-6-methylisonicotinic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.