missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-4-(trifluoromethyl)phenol, 97%, Thermo Scientific™

60.58 EUR

Chemical Identifiers

CAS 463976-21-8
Molecular Formula C7H4F3IO
Molecular Weight (g/mol) 288.008
MDL Number MFCD12922960
InChI Key MRDCXUYQUSFQKF-UHFFFAOYSA-N
Synonym 2-iodo-4-trifluoromethyl phenol, phenol, 2-iodo-4-trifluoromethyl, 4-hydroxy-3-iodobenzotrifluoride, pubchem20185, 4-trifluoromethyl-6-iodophenol, 2-iodo-4-trifluoromethyl-phenol, 4-trifluoromethyl-2-iodophenol, alpha,alpha,alpha-trifluoro-o-iodo-p-cresol
PubChem CID 11572816
IUPAC Name 2-iodo-4-(trifluoromethyl)phenol
SMILES C1=CC(=C(C=C1C(F)(F)F)I)O
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15446817
SDS View Documents
Thermo Scientific Alfa Aesar
H33928.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Iodo-4-(trifluoromethyl)phenol is an pharmaceutical intermediate and it is an intermediate in organic synthesis.

Notes
Light sensitive. Store in the dark. Store away from oxidizing agent, light.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Iodo-4-(trifluoromethyl)phenol, 97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.