Learn More
2-Isopropylaniline, 97%, Thermo Scientific Chemicals Product Code.: 11457646
2-Isopropylaniline, 97%, Thermo Scientific Chemicals
Product Code. 11457646
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
Product Code. 11457646
Brand: Thermo Scientific Alfa Aesar A14671.22
Request Bulk or Custom Format
2-Isopropylaniline is used in the synthesis of a colorless amine-coordinated zinc complex. The reactions of 1,1?-bis(hydroxymethyl)ferrocene with 2-isopropylaniline, in the presence of a catalyst [RuCl2(PPh3)3], was studied
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Isopropylaniline is used in the synthesis of a colorless amine-coordinated zinc complex. The reactions of 1,1′-bis(hydroxymethyl)ferrocene with 2-isopropylaniline, in the presence of a catalyst [RuCl2(PPh3)3], was studied
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong oxidizing agents, strong acids, acid chlorides, acid anhydrides. Light Sensitive
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 643-28-7
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00007720
InChI Key YKOLZVXSPGIIBJ-UHFFFAOYSA-N
Synonym 2-isopropylaniline, o-isopropylaniline, o-cumidine, 2-propan-2-yl aniline, benzenamine, 2-1-methylethyl, o-aminoisopropylbenzene, 2-isopropyl aniline, 2-aminoisopropylbenzene, aniline, o-isopropyl, o-isopropyl aniline
PubChem CID 12561
SMILES CC(C)C1=CC=CC=C1N
Specifications
Density 0.966
Boiling Point 214°C to 216°C
Flash Point 95°C (203°F)
Refractive Index 1.5485
Quantity 100 g
Beilstein 636283
Sensitivity Light sensitive
Solubility Information Insoluble in water.
IUPAC Name 2-propan-2-ylaniline
Formula Weight 135.21
Percent Purity 97%
Chemical Name or Material 2-Isopropylaniline
Product Title
2-Isopropylaniline, 97%, Thermo Scientific Chemicals >