Learn More
2-Isopropylanisole, 98%, Thermo Scientific Chemicals
Description
2-Isopropylanisole is the startind material in thesynthesis of (±)-shonanyl methyl ether and (±)-ferruginyl methyl ether.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Isopropylanisole is the startind material in thesynthesis of (±)-shonanyl methyl ether and (±)-ferruginyl methyl ether.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Specifications
Specifications
Density | 0.936 |
Boiling Point | 198°C to 200°C |
Flash Point | 86°C (187°F) |
Refractive Index | 1.508 |
Quantity | 25 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 150.22 |
Percent Purity | 98% |
Chemical Name or Material | 2-Isopropylanisole |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.