Learn More
2-Methylbutyric acid, 98%, Thermo Scientific Chemicals
26.63 EUR - 573.86 EUR
Chemical Identifiers
CAS | 116-53-0 |
---|---|
Molecular Formula | C5H10O2 |
Molecular Weight (g/mol) | 102.133 |
MDL Number | MFCD00002669 |
InChI Key | WLAMNBDJUVNPJU-UHFFFAOYSA-N |
Synonym | 2-methylbutyric acid, dl-2-methylbutyric acid, butanoic acid, 2-methyl, ethylmethylacetic acid, methylbutyric acid, 2-methyl butyric acid, 2-methybutyric acid, methylethylacetic acid, active valeric acid, butyric acid, 2-methyl |
PubChem CID | 8314 |
ChEBI | CHEBI:37070 |
IUPAC Name | 2-methylbutanoic acid |
SMILES | CCC(C)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11349579
|
Thermo Scientific Alfa Aesar
A11546.AC |
25 mL |
26.63 EUR
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11444573
|
Thermo Scientific Alfa Aesar
A11546.AE |
100 mL |
37.69 EUR
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11454573
|
Thermo Scientific Alfa Aesar
A11546.AP |
500 mL |
133.67 EUR
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11464573
|
Thermo Scientific Alfa Aesar
A11546.0F |
2500 mL |
573.86 EUR
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Methylbutyric acid, can be used as food additive for butter, cream, cheese flavor deployment. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used in the synthesis of 2-methylbutyric anhydride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Methylbutyric acid, can be used as food additive for butter, cream, cheese flavor deployment. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used in the synthesis of 2-methylbutyric anhydride.
Solubility
It is soluble in water, but highly soluble in most common organic solvents such as Chloroform, Methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Bases, Oxidizing agents, Reducing agents.
Chemical Identifiers
116-53-0 | |
102.133 | |
WLAMNBDJUVNPJU-UHFFFAOYSA-N | |
8314 | |
2-methylbutanoic acid |
C5H10O2 | |
MFCD00002669 | |
2-methylbutyric acid, dl-2-methylbutyric acid, butanoic acid, 2-methyl, ethylmethylacetic acid, methylbutyric acid, 2-methyl butyric acid, 2-methybutyric acid, methylethylacetic acid, active valeric acid, butyric acid, 2-methyl | |
CHEBI:37070 | |
CCC(C)C(=O)O |
Specifications
116-53-0 | |
175°C to 176°C | |
98% | |
1.4055 | |
UN3265 | |
2-methylbutyric acid, dl-2-methylbutyric acid, butanoic acid, 2-methyl, ethylmethylacetic acid, methylbutyric acid, 2-methyl butyric acid, 2-methybutyric acid, methylethylacetic acid, active valeric acid, butyric acid, 2-methyl | |
WLAMNBDJUVNPJU-UHFFFAOYSA-N | |
2-methylbutanoic acid | |
8314 | |
102.14 | |
Liquid |
0.937 | |
73°C (163°F) | |
C5H10O2 | |
MFCD00002669 | |
1720486 | |
It is soluble in water,but highly soluble in most common organic solvents such as Chloroform,Methanol. | |
CCC(C)C(=O)O | |
102.133 | |
CHEBI:37070 | |
98% | |
2-Methylbutyric acid |
Safety and Handling
GHS H Statement
H301-H311-H314-H318-H227
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302+H312-H314
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 204-145-2
missing translation for 'rtecsNumber' : EK7897000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- 2-Methylbutyric acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : 2-Methylbutyric acid
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.