Learn More
2-Thiophenemethylamine, 94%, Thermo Scientific Chemicals
50.45 EUR - 460.29 EUR
Chemical Identifiers
CAS | 27757-85-3 |
---|---|
Molecular Formula | C5H7NS |
Molecular Weight (g/mol) | 113.178 |
MDL Number | MFCD00005460 |
InChI Key | FKKJJPMGAWGYPN-UHFFFAOYSA-N |
Synonym | 2-thiophenemethylamine, 2-thienylmethylamine, 2-thiophenemethanamine, 2-aminomethyl thiophene, thiophene-2-methylamine, thiophenemethanamine, 2-aminomethylthiophene, 2-thenylamine, 2-thienylmethyl amine, 1-thiophen-2-yl methanamine |
PubChem CID | 34005 |
IUPAC Name | thiophen-2-ylmethanamine |
SMILES | C1=CSC(=C1)CN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11439213
|
Thermo Scientific Alfa Aesar
A11700.06 |
5 g |
50.45 EUR
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11449213
|
Thermo Scientific Alfa Aesar
A11700.14 |
25 g |
164.82 EUR
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11459213
|
Thermo Scientific Alfa Aesar
A11700.22 |
100 g |
460.29 EUR
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Thiophenemethylamine is Reactant involved in synthesis of: Triazole-linked-thiopene conjugates. It is used for biomimetic model for studies of metal detoxification and oxidative stress involving metallothionein.It is a reactant involved in studies of organocatalyzed asymmetric reductive amination of ketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Thiophenemethylamine is Reactant involved in synthesis of: Triazole-linked-thiopene conjugates. It is used for biomimetic model for studies of metal detoxification and oxidative stress involving metallothionein.It is a reactant involved in studies of organocatalyzed asymmetric reductive amination of ketones.
Solubility
Easily soluble in ethanol and Chloroform.
Notes
Air Sensitive. Store under dry inert gas. Store away from air.
Chemical Identifiers
27757-85-3 | |
113.178 | |
FKKJJPMGAWGYPN-UHFFFAOYSA-N | |
34005 | |
C1=CSC(=C1)CN |
C5H7NS | |
MFCD00005460 | |
2-thiophenemethylamine, 2-thienylmethylamine, 2-thiophenemethanamine, 2-aminomethyl thiophene, thiophene-2-methylamine, thiophenemethanamine, 2-aminomethylthiophene, 2-thenylamine, 2-thienylmethyl amine, 1-thiophen-2-yl methanamine | |
thiophen-2-ylmethanamine |
Specifications
27757-85-3 | |
85°C to 88°C (14 mmHg) | |
C5H7NS | |
MFCD00005460 | |
106296 | |
2-thiophenemethylamine, 2-thienylmethylamine, 2-thiophenemethanamine, 2-aminomethyl thiophene, thiophene-2-methylamine, thiophenemethanamine, 2-aminomethylthiophene, 2-thenylamine, 2-thienylmethyl amine, 1-thiophen-2-yl methanamine | |
FKKJJPMGAWGYPN-UHFFFAOYSA-N | |
thiophen-2-ylmethanamine | |
34005 | |
97% |
1.1 | |
73°C (163°F) | |
1.567 | |
UN2735 | |
Air Sensitive | |
Easily soluble in ethanol and Chloroform. | |
C1=CSC(=C1)CN | |
113.178 | |
113.19 | |
2-Thiophenemethylamine |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 248-639-6
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.