Learn More
20-Deoxyingenol, MedChemExpress
MedChemExpress 20-Deoxyingenol, a diterpene, is isolated from the roots of Euphorbia kansui. 20-Deoxyingenol can promote autophagy and lysosomal biogenesis by promoting the nuclear translocation of transcription factor EB (TFEB) in vitro. 20-Deoxyingenol can be used for the research of osteoarthritis (OA).
Brand:  Medchem Express HY-N0866
Encompass_Preferred
 View more versions of this product
Product Code. 16421558
  • 167.40 EUR / 5mg
Explore more special offers
Description
Description
20-Deoxyingenol, a diterpene, is isolated from the roots of Euphorbia kansui. 20-Deoxyingenol can promote autophagy and lysosomal biogenesis by promoting the nuclear translocation of transcription factor EB (TFEB) in vitro. 20-Deoxyingenol can be used for the research of osteoarthritis (OA).
Specifications
Specifications
20-Deoxyingenol
White
C20H28O4
DMSO : 50 mg/mL (150.41 mM; Need ultrasonic)
332.43
98.0%
Solid
54706-99-9
Research
5 mg
O=C1[C@@]2(C=C(C)[C@@H]3O)[C@]3(O)[C@H](O)C(C)=C[C@@]1([H])[C@@]4([H])[C@@](C4(C)C)([H])C[C@H]2C
332.43
Research
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
20-Deoxyingenol, MedChemExpress