Learn More
3M™ 2890 Series Polycarbonate Lens Safety Goggles
Brand:  3M™ 7000032483
13.50 EUR valid until 2025-06-08
Use promo code "24131" to get your promotional price.
 View more versions of this product
Product Code. 12672605
  • 26.00 EUR / Each
Estimated Shipment: 23-04-2025
Specifications
EN 166:2001
Clear
Construction,‎ laboratories
Sealed
Anti-fog
Safety Goggles
1 Ea.
Yes
Special Offers
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131
    Expires2025-06-08
    New Offer
    Shop Now
Product Content Correction
Product Title
3M™ 2890 Series Polycarbonate Lens Safety Goggles > Lens Tint: Clear; Lens Coating: Anti-fog