missing translation for 'onlineSavingsMsg'
Learn More
3-[2-Methyl-5-(trifluoromethoxy)phenyl]propionic acid, 97%, Thermo Scientific™ Product Code.: 15467257

3-[2-Methyl-5-(trifluoromethoxy)phenyl]propionic acid, 97%, Thermo Scientific™

Product Code. 15467257
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15467257

Brand: Thermo Scientific Alfa Aesar H34318.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1373921-10-8
Molecular Formula C11H11F3O3
Molecular Weight (g/mol) 248.201
MDL Number MFCD22201057
InChI Key HRXITAZJWHWLCF-UHFFFAOYSA-N
Synonym 3-2-methyl-5-trifluoromethoxy phenyl propionic acid, 3-2-methyl-5-trifluoromethoxy phenyl propanoic acid, 3-2-methyl-5-trifluoromethoxy-phenyl propionic acid
PubChem CID 86277663
IUPAC Name 3-[2-methyl-5-(trifluoromethoxy)phenyl]propanoic acid
SMILES CC1=C(C=C(C=C1)OC(F)(F)F)CCC(=O)O

Specifications

Quantity 1 g
Formula Weight 248.2
Percent Purity 97%
Chemical Name or Material 3-[2-Methyl-5-(trifluoromethoxy)phenyl]propionic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-[2-Methyl-5-(trifluoromethoxy)phenyl]propionic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.