Learn More
3,3',4,4'-Benzophenonetetracarboxylic dianhydride, 97+%, Thermo Scientific Chemicals
Description
BTDA polymer fine free-flowing powder suitable for general purposes. BTDA Microfine Micronized powder facilitates mixing in epoxy resins. BTDA ultrapure high-purity, high-assay powder for critical polyimide synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
BTDA polymer fine free-flowing powder suitable for general purposes. BTDA Microfine Micronized powder facilitates mixing in epoxy resins. BTDA ultrapure high-purity, high-assay powder for critical polyimide synthesis.
Solubility
Soluble in DMF (0.1 g/mL). Reacts with water.
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Specifications
Specifications
Melting Point | 220°C to 226°C |
Density | 1.57 |
Boiling Point | 320°C (5 mmHg) |
Flash Point | 324°C (615°F) |
Quantity | 2.5 kg |
Beilstein | 761777 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in DMF (0.1g/mL). Reacts with water. |
Formula Weight | 322.23 |
Percent Purity | ≥97% |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
U21J008 | Certificate of Analysis | 05/09/2023 | A16529.36,A16529.A4,A16529.22 |
10244206 | Certificate of Analysis | 07/08/2023 | A16529.36,A16529.A4,A16529.22 |
P11I040 | Certificate of Analysis | 20/06/2022 | A16529.36,A16529.A4,A16529.22 |
X26H060 | Certificate of Analysis | 12/04/2022 | A16529.36,A16529.A4,A16529.22 |
W08H038 | Certificate of Analysis | 12/04/2022 | A16529.36,A16529.A4,A16529.22 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.