missing translation for 'onlineSavingsMsg'
Learn More
3,3',4,4'-Benzophenonetetracarboxylic dianhydride, 97+%, Thermo Scientific Chemicals Product Code.: 11457857

3,3',4,4'-Benzophenonetetracarboxylic dianhydride, 97+%, Thermo Scientific Chemicals

Product Code. 11457857
100 g
Click to view available options
Quantity:
100 g
500 g
2.5 kg
This item is not returnable. View return policy

Product Code. 11457857

Brand: Thermo Scientific Alfa Aesar A16529.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

BTDA polymer fine free-flowing powder suitable for general purposes. BTDA Microfine Micronized powder facilitates mixing in epoxy resins. BTDA ultrapure high-purity, high-assay powder for critical polyimide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
BTDA polymer fine free-flowing powder suitable for general purposes. BTDA Microfine Micronized powder facilitates mixing in epoxy resins. BTDA ultrapure high-purity, high-assay powder for critical polyimide synthesis.

Solubility
Soluble in DMF (0.1 g/mL). Reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2421-28-5
Molecular Formula C17H6O7
Molecular Weight (g/mol) 322.228
MDL Number MFCD00005923
InChI Key VQVIHDPBMFABCQ-UHFFFAOYSA-N
Synonym 3,3',4,4'-benzophenonetetracarboxylic dianhydride, benzophenonetetracarboxylic dianhydride, benzophenonetetracarboxylic acid dianhydride, 1,3-isobenzofurandione, 5,5'-carbonylbis, 4,4'-carbonylbis phthalic anhydride, 4,4'-carbonyldiphthalic anhydride, 4,4'-diphthalic anhydride ketone, benzophenonetetracarboxylic anhydride, phthalic anhydride, 4,4'-carbonyldi, unii-y61gva8097Show MoreShow Less
PubChem CID 75498
IUPAC Name 5-(1,3-dioxo-2-benzofuran-5-carbonyl)-2-benzofuran-1,3-dione
SMILES C1=CC2=C(C=C1C(=O)C3=CC4=C(C=C3)C(=O)OC4=O)C(=O)OC2=O

Specifications

Melting Point 220°C to 226°C
Density 1.57
Boiling Point 320°C (5 mmHg)
Flash Point 324°C (615°F)
Quantity 100 g
Beilstein 761777
Sensitivity Moisture sensitive
Solubility Information Soluble in DMF (0.1g/mL). Reacts with water.
Formula Weight 322.23
Percent Purity ≥97%
Chemical Name or Material 3,3',4,4'-Benzophenonetetracarboxylic dianhydride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU21J008 Certificate TypeCertificate of Analysis Date05/09/2023 Catalog NumberA16529.36,A16529.A4,A16529.22
Lot Number10244206 Certificate TypeCertificate of Analysis Date07/08/2023 Catalog NumberA16529.36,A16529.A4,A16529.22
Lot NumberP11I040 Certificate TypeCertificate of Analysis Date20/06/2022 Catalog NumberA16529.36,A16529.A4,A16529.22
Lot NumberX26H060 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA16529.36,A16529.A4,A16529.22
Lot NumberW08H038 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA16529.36,A16529.A4,A16529.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,3',4,4'-Benzophenonetetracarboxylic dianhydride, 97+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.