missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Methoxyphenyl)propionic acid, 99%, Thermo Scientific™

44.50 EUR - 141.00 EUR

Chemical Identifiers

CAS 10516-71-9
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00014027
InChI Key BJJQJLOZWBZEGA-UHFFFAOYSA-N
Synonym 3-3-methoxyphenyl propionic acid, 3-3-methoxyphenyl propanoic acid, 3-m-methoxyphenyl propionic acid, 3-methoxybenzenepropanoic acid, benzenepropanoic acid, 3-methoxy, 3-3-methoxy-phenyl-propionic acid, 3-methoxy-benzenepropanoic acid, 3-3-methoxyphenyl propanoate, m-methoxyhydrocinnamate, 3-methoxydihydrocinnamate
PubChem CID 66336
IUPAC Name 3-(3-methoxyphenyl)propanoic acid
SMILES COC1=CC=CC(CCC(O)=O)=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15432212
View Documents
Thermo Scientific Acros
458110050
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15442212
View Documents
Thermo Scientific Acros
458110250
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 234-049-6

Product Identifier
  • 3-(3-Methoxyphenyl)propionic acid

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-(3-Methoxyphenyl)propionic acid, 99%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.