missing translation for 'onlineSavingsMsg'
Learn More
3',4',5,7-Tetramethoxyflavone, 97%, Thermo Scientific Chemicals Product Code.: 11313556

3',4',5,7-Tetramethoxyflavone, 97%, Thermo Scientific Chemicals

Product Code. 11313556
500 mg
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy

Product Code. 11313556

Brand: Thermo Scientific Alfa Aesar L14496.ME

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 855-97-0
Molecular Formula C19H18O6
Molecular Weight (g/mol) 342.35
MDL Number MFCD00017558
InChI Key CLXVBVLQKLQNRQ-UHFFFAOYSA-N
Synonym 3',4',5,7-tetramethoxyflavone, luteolin tetramethyl ether, tetramethoxyluteolin, 5,7,3',4'-tetramethylluteolin, tetramethylluteolin, 2-3,4-dimethoxyphenyl-5,7-dimethoxy-4h-chromen-4-one, luteolin tetramethylether, 2-3,4-dimethoxyphenyl-5,7-dimethoxychromen-4-one, luteolin 5,7,3',4'-tetramethyl ether, 4h-1-benzopyran-4-one, 2-3,4-dimethoxyphenyl-5,7-dimethoxyShow MoreShow Less
PubChem CID 631170
IUPAC Name 2-(3,4-dimethoxyphenyl)-5,7-dimethoxychromen-4-one
SMILES COC1=CC(OC)=C2C(=O)C=C(OC2=C1)C1=CC=C(OC)C(OC)=C1

Specifications

Melting Point 193°C to 197°C
Quantity 500 mg
Beilstein 336320
Formula Weight 342.35
Percent Purity 97%
Chemical Name or Material 3',4',5,7-Tetramethoxyflavone

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250412 Certificate TypeCertificate of Analysis Date11/04/2024 Catalog NumberL14496.ME,L14496.MC
Lot Number10227215 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL14496.ME,L14496.MC
Lot NumberGBFA011272 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL14496.ME,L14496.MC
Lot Number10108511 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL14496.ME,L14496.MC

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3',4',5,7-Tetramethoxyflavone, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.