missing translation for 'onlineSavingsMsg'
Learn More
3,4,5-Trichloropyridine, 98%, Thermo Scientific Chemicals Product Code.: 11434304

3,4,5-Trichloropyridine, 98%, Thermo Scientific Chemicals

Product Code. 11434304
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11434304

Brand: Thermo Scientific Alfa Aesar A12587.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33216-52-3
Molecular Formula C5H2Cl3N
Molecular Weight (g/mol) 182.428
MDL Number MFCD00051685
InChI Key KKWRVUBDCJQHBZ-UHFFFAOYSA-N
Synonym pyridine, 3,4,5-trichloro, 3,4,5-trichloro-pyridine, pubchem11021, acmc-209hzc, ksc497k5t, 3,4,5-trichloropyridine, 3,4,5-trichloro pyridine, 3,4,5-trichloropyridine 1gShow MoreShow Less
PubChem CID 3786531
IUPAC Name 3,4,5-trichloropyridine
SMILES C1=C(C(=C(C=N1)Cl)Cl)Cl

Specifications

Melting Point 70°C to 74°C
Boiling Point 213°C to 215°C
Quantity 1 g
Beilstein 120542
Formula Weight 182.44
Percent Purity 98%
Chemical Name or Material 3,4,5-Trichloropyridine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252448 Certificate TypeCertificate of Analysis Date17/09/2024 Catalog NumberA12587.06,A12587.03
Lot Number10242248 Certificate TypeCertificate of Analysis Date22/07/2022 Catalog NumberA12587.06,A12587.03,A12587.14
Lot Number10114404 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12587.06,A12587.03,A12587.14
Lot Number10140221 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12587.06,A12587.03,A12587.14
Lot Number10222872 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12587.06,A12587.03,A12587.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,4,5-Trichloropyridine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.