missing translation for 'onlineSavingsMsg'
Learn More
3,4,5-Trimethoxyphenylacetic acid, 99%, Thermo Scientific Chemicals Product Code.: 11424627

3,4,5-Trimethoxyphenylacetic acid, 99%, Thermo Scientific Chemicals

Product Code. 11424627
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11424627

Brand: Thermo Scientific Alfa Aesar B21136.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used in the synthesis of 3,4-Diaryl-2(5H)-furanones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of 3,4-Diaryl-2(5H)-furanones.

Solubility
Soluble in water. Solubility in methanol is almost transparent.

Notes
Store away from bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 951-82-6
Molecular Formula C11H14O5
Molecular Weight (g/mol) 226.228
MDL Number MFCD00004336
InChI Key DDSJXCGGOXKGSJ-UHFFFAOYSA-N
Synonym 3,4,5-trimethoxyphenylacetic acid, 2-3,4,5-trimethoxyphenyl acetic acid, 3,4,5-trimethoxyphenyl acetic acid, benzeneacetic acid, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenylaceticacid, pubchem14015, acmc-209rv8, ksc487g4j, 3,4,5-trimethoxybenzeneacetic acid, benzeneacetic acid,4,5-trimethoxyShow MoreShow Less
PubChem CID 70372
IUPAC Name 2-(3,4,5-trimethoxyphenyl)acetic acid
SMILES COC1=CC(=CC(=C1OC)OC)CC(=O)O

Specifications

Melting Point 118°C to 122°C
Quantity 25 g
Beilstein 2697844
Solubility Information Soluble in water. Solubility in methanol is almost transparent.
Formula Weight 226.23
Percent Purity 99%
Chemical Name or Material 3,4,5-Trimethoxyphenylacetic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10238514 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberB21136.06,B21136.14,B21136.22
Lot NumberJ6869C Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21136.06,B21136.14,B21136.22
Lot Number10190994 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21136.06,B21136.14,B21136.22
Lot Number10159557 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21136.06,B21136.14,B21136.22
Lot Number10177003 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21136.06,B21136.14,B21136.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,4,5-Trimethoxyphenylacetic acid, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.