Learn More
3,5-Bis(trifluoromethyl)aniline, 98+%, Thermo Scientific Chemicals
33.97 EUR - 431.00 EUR
Chemical Identifiers
CAS | 328-74-5 |
---|---|
Molecular Formula | C8H5F6N |
Molecular Weight (g/mol) | 229.12 |
MDL Number | MFCD00000394 |
InChI Key | CDIDGWDGQGVCIB-UHFFFAOYSA-N |
Synonym | 3,5-bis trifluoromethyl aniline, 3,5-di trifluoromethyl aniline, benzenamine, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzenamine, 3',5'-bis-trifluoromethyl aniline, 3,5-bis-trifluoromethylaniline, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-3,5-xylidine, 3,5-bis-trifluoromethyl-phenylamine, aniline, 3,5-bis trifluoromethyl |
PubChem CID | 9480 |
IUPAC Name | 3,5-bis(trifluoromethyl)aniline |
SMILES | C1=C(C=C(C=C1C(F)(F)F)N)C(F)(F)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
10357220
|
Thermo Scientific Acros
185690100 |
10 mL | Glass bottle |
33.97 EUR
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
10721081
|
Thermo Scientific Acros
185690500 |
50 mL | Glass bottle |
119.60 EUR
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
10453965
|
Thermo Scientific Acros
185692500 |
250 mL | Glass bottle | N/A | N/A | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
328-74-5 | |
229.12 | |
CDIDGWDGQGVCIB-UHFFFAOYSA-N | |
9480 | |
C1=C(C=C(C=C1C(F)(F)F)N)C(F)(F)F |
C8H5F6N | |
MFCD00000394 | |
3,5-bis trifluoromethyl aniline, 3,5-di trifluoromethyl aniline, benzenamine, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzenamine, 3',5'-bis-trifluoromethyl aniline, 3,5-bis-trifluoromethylaniline, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-3,5-xylidine, 3,5-bis-trifluoromethyl-phenylamine, aniline, 3,5-bis trifluoromethyl | |
3,5-bis(trifluoromethyl)aniline |
Specifications
328-74-5 | |
100.0 | |
1.4700g/mL | |
83°C | |
98+% | |
(CF3)2C6H3NH2 | |
MFCD00000394 | |
3,5-bis trifluoromethyl aniline, 3,5-di trifluoromethyl aniline, benzenamine, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzenamine, 3',5'-bis-trifluoromethyl aniline, 3,5-bis-trifluoromethylaniline, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-3,5-xylidine, 3,5-bis-trifluoromethyl-phenylamine, aniline, 3,5-bis trifluoromethyl | |
C1=C(C=C(C=C1C(F)(F)F)N)C(F)(F)F | |
229.12 | |
229.12 | |
Liquid |
98.0 | |
Brown-Yellow to Yellow | |
85°C (15.0 mmHg) | |
Authentic | |
C8H5F6N | |
1.4330 to 1.435 | |
1.47 | |
CDIDGWDGQGVCIB-UHFFFAOYSA-N | |
3,5-bis(trifluoromethyl)aniline | |
9480 | |
98+% | |
3, 5-Bis(trifluoromethyl)aniline |
Safety and Handling
GHS H Statement
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remov
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 206-335-
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.